Learn More
Barium silicon oxide, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01512236
Description
Barium silicate films that are suitable for use as transmission media in integrated optical circuits. It is shown that the films, which can be produced with a wide range of refractive indices by suitable selection of the ratio of the target constituents, exhibit low optical attenuation. Hence it is used for integrated optical circuits. It is used in ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Barium silicon oxide | |
| White | |
| Powder | |
| BaO3Si | |
| barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1 | |
| HMOQPOVBDRFNIU-UHFFFAOYSA-N | |
| barium(2+);dioxido(oxo)silane | |
| 166725 | |
| 1.67 |
| 1604°C | |
| 13255-26-0 | |
| 500 g | |
| MFCD00014186 | |
| Insoluble in water. Soluble in acids. | |
| [O-][Si](=O)[O-].[Ba+2] | |
| 213.409 | |
| 213.4 | |
| 4.399 |
Chemical Identifiers
| 13255-26-0 | |
| 213.409 | |
| HMOQPOVBDRFNIU-UHFFFAOYSA-N | |
| 166725 | |
| [O-][Si](=O)[O-].[Ba+2] |
| BaO3Si | |
| MFCD00014186 | |
| barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1 | |
| barium(2+);dioxido(oxo)silane |
Safety and Handling
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : DOT Class: 6.1, Packaging Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only