accessibility menu, dialog, popup

UserName

CAS RN 13255-26-0

BaOSiOO

CAS RN 13255-26-0

IUPAC Name: barium(2+) oxosilanebis(olate)
Synonyms: barium(2+) oxosilanebis(olate)
Molecular Weight (g/mol): 213.41
Molecular Formula: BaO3Si
InChi Key: HMOQPOVBDRFNIU-UHFFFAOYSA-N
SMILES: [Ba++].[O-][Si]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results