missing translation for 'onlineSavingsMsg'
Learn More

Barium silicon oxide, Thermo Scientific Chemicals

$430.74 - $430.74

Chemical Identifiers

CAS 13255-26-0
Molecular Formula BaO3Si
Molecular Weight (g/mol) 213.409
MDL Number MFCD00014186
InChI Key HMOQPOVBDRFNIU-UHFFFAOYSA-N
Synonym barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1
PubChem CID 166725
IUPAC Name barium(2+);dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[Ba+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1512236
View Documents
Thermo Scientific Chemicals
01512236
500 g
Each for $430.74
Only null left
 
Description

Description

Barium silicate films that are suitable for use as transmission media in integrated optical circuits. It is shown that the films, which can be produced with a wide range of refractive indices by suitable selection of the ratio of the target constituents, exhibit low optical attenuation. Hence it is used for integrated optical circuits. It is used in ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13255-26-0
213.409
HMOQPOVBDRFNIU-UHFFFAOYSA-N
166725
[O-][Si](=O)[O-].[Ba+2]
BaO3Si
MFCD00014186
barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1
barium(2+);dioxido(oxo)silane

Specifications

1604°C
13255-26-0
500 g
MFCD00014186
Insoluble in water. Soluble in acids.
[O-][Si](=O)[O-].[Ba+2]
213.409
213.4
4.399
White
Powder
BaO3Si
barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1
HMOQPOVBDRFNIU-UHFFFAOYSA-N
barium(2+);dioxido(oxo)silane
166725
1.67
Barium silicon oxide
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

DOTInformation : DOT Class: 6.1, Packaging Group: III

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only