missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide TS, Saturated, pH 12.9, Ricca Chemical

Catalog No. 89832
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89832 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89832 Supplier Ricca Chemical Company Supplier No. 89832

Chemical Identifiers

CAS 17194-00-2 , 7732-18-5
Molecular Formula BaH2O2
Molecular Weight (g/mol) 171.341
InChI Key RQPZNWPYLFFXCP-UHFFFAOYSA-L
Synonym barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous
PubChem CID 6093286
ChEBI CHEBI:32592
IUPAC Name barium(2+);dihydroxide
SMILES [OH-].[OH-].[Ba+2]

Specifications

Color Colorless
CAS Min % 96.0, 2.39
CAS Max % 100.0, 2.49
Grade Laboratory
Packaging Natural Poly Bottle
pH 12.9
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes Saturated Aqueous Solution
Chemical Name or Material Barium Hydroxide, Test Solution, pH 12.9