missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride TS/RS, 12% (w/v), Ricca Chemical
$146.21 - $210.80
Chemical Identifiers
| CAS | 10326-27-9 |
|---|---|
| Molecular Formula | BaCl2H4O2 |
| Molecular Weight (g/mol) | 244.26 |
| MDL Number | MFCD00149154 |
| InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
| Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
| PubChem CID | 5284346 |
| ChEBI | CHEBI:86153 |
| IUPAC Name | barium(2+) dihydrate dichloride |
| SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Chemical Identifiers
| 10326-27-9 | |
| 244.26 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| 5284346 | |
| barium(2+) dihydrate dichloride |
| BaCl2H4O2 | |
| MFCD00149154 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| CHEBI:86153 | |
| O.O.[Cl-].[Cl-].[Ba++] |
Specifications
| 10326-27-9 , 7732-18-5 | |
| 91.0,10.93 | |
| MFCD00149154 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+) dihydrate dichloride | |
| 5284346 | |
| 12% | |
| Natural Poly Bottle | |
| 500 mL |
| 87.0,10.51 | |
| BaCl2H4O2 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| O.O.[Cl-].[Cl-].[Ba++] | |
| 244.26 | |
| CHEBI:86153 | |
| Laboratory | |
| ∼5 to 7 | |
| Barium Chloride (12% w/v) |