missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, For Sulfate, Certified, 10.0% (w/v) ±0.5%, LabChem™

Catalog No. LC116005
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC116005 20 L
LC116001 500 mL
LC116002 1 L
LC116004 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC116005 Supplier LabChem Supplier No. LC116005
Only null left

Chemical Identifiers

CAS 10326-27-9 , 7732-18-5
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Color Colorless
CAS Max % 90.78, 9.22
Linear Formula BaCl2·2H2O
Density 1.08g/mL
Solubility Information Soluble in water
Formula Weight 244.26
Percent Purity 10.0% ±0.5% (w/v)
Grade Certified
Identification Passes Test
Packaging Cubitainer™
Specific Gravity 1.08g/mL
Traceability to NIST Traceable to NIST
Viscosity 1.03cSt
Decomposition Information Hydrogen chloride; Barium
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Barium Chloride, For Sulfate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature