missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, For Sulfate, Certified, 10.0% (w/v) ±0.5%, LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 90.78, 9.22 |
| Linear Formula | BaCl2·2H2O |
| Density | 1.08g/mL |
| Solubility Information | Soluble in water |
| Formula Weight | 244.26 |
| Percent Purity | 10.0% ±0.5% (w/v) |
| Grade | Certified |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Wash exposed skin thoroughly after handling. Do not eat, drink or smoke when using this product. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |