missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 33% (w/v), Ricca Chemical

Numéro de catalogue. 8685
Modifier l'affichage
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
8685 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 8685 Fournisseur Ricca Chemical Company Code fournisseur 8685
Il en reste null

Identifiants chimiques

CAS 10326-27-9 , 7732-18-5
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Spécifications

Color Colorless
CAS Min % 74.0, 32.0
CAS Max % 77.0, 34.0
Percent Purity 33%
Grade Laboratory
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
pH ∼5 to 7
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 33% (w/v)
Chemical Name or Material Barium Chloride
Afficher plus de résultats Afficher moins de résultats