missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 22.5°C Baumé, Ricca Chemical

Supplier:  Ricca Chemical Company R08700004A

Quantity 4 L
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. R08700004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

10326-27-9 , 7732-18-5
244.257
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba+2]
BaCl2H4O2
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+);dichloride;dihydrate

Specifications

Barium Chloride
10326-27-9 , 7732-18-5
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
O.O.[Cl-].[Cl-].[Ba+2]
244.257
CHEBI:86153
4 L
22.5°F Baumæ at 60°
BaCl2H4O2
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+);dichloride;dihydrate
5284346
Natural Poly Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents

Documents