missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 20% (w/v), Ricca Chemical

$97.63 - $398.50

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
855-16
SDS View Documents
Ricca Chemical Company
85516
500 mL Natural Poly Bottle
Each for $97.63
Only null left
 
855-32
View Documents
Ricca Chemical Company
RI85532
1 L Natural Poly Bottle
Each for $159.60
Only null left
 
855-1
SDS View Documents
Ricca Chemical Company
8551
4 L Natural Poly Bottle
Each for $398.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+) dihydrate dichloride
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba++]

Specifications

Colorless
82.0,19.0
BaCl2H4O2
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
Laboratory
Aqueous Solution
500 mL
20% (w/v)
10326-27-9 , 7732-18-5
85.0,21.0
MFCD00149154
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
20%
Natural Poly Bottle
∼5 to 7
Liquid
Barium Chloride
Videos
SDS
Documents

Documents