missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride, 11.8% (w/v) (118 g/L), Ricca Chemical

Supplier:  Ricca Chemical Company 85232

 View more versions of this product

Catalog No. 852-32

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Barium Chloride
88.0,11.0
BaCl2H4O2
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+);dichloride;dihydrate
5284346
Natural Poly Bottle
1 L
10326-27-9 , 7732-18-5
91.0,12.0
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
O.O.[Cl-].[Cl-].[Ba+2]
244.257
CHEBI:86153
Aqueous Solution
11.8% (w/v); 118g/L

Chemical Identifiers

10326-27-9
244.257
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba+2]
BaCl2H4O2
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+);dichloride;dihydrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions