missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, 11.8% (w/v) (118 g/L), Ricca Chemical
Chemical Identifiers
| CAS | 10326-27-9 |
|---|---|
| Molecular Formula | BaCl2H4O2 |
| Molecular Weight (g/mol) | 244.257 |
| InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
| Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
| PubChem CID | 5284346 |
| ChEBI | CHEBI:86153 |
| IUPAC Name | barium(2+);dichloride;dihydrate |
| SMILES | O.O.[Cl-].[Cl-].[Ba+2] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
852-16
|
Ricca Chemical Company
85216 |
500 mL | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
852-32
|
Ricca Chemical Company
85232 |
1 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 10326-27-9 | |
| 244.257 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| CHEBI:86153 | |
| O.O.[Cl-].[Cl-].[Ba+2] |
| BaCl2H4O2 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| 5284346 | |
| barium(2+);dichloride;dihydrate |
Specifications
| 10326-27-9 , 7732-18-5 | |
| 91.0,12.0 | |
| barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
| O.O.[Cl-].[Cl-].[Ba+2] | |
| 244.257 | |
| CHEBI:86153 | |
| Aqueous Solution | |
| 11.8% (w/v); 118g/L |
| 88.0,11.0 | |
| BaCl2H4O2 | |
| PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
| barium(2+);dichloride;dihydrate | |
| 5284346 | |
| Natural Poly Bottle | |
| 500 mL | |
| Barium Chloride |