missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, 10% (w/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 89.0, 9.0 |
| CAS Max % | 93.0, 11.0 |
| Percent Purity | 10% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| pH | 6.5 to 6.9 |
| Quantity | 4 L |
| Physical Form | Liquid |
| Show More |