missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium AA Standard, Certified, 1000ppm Ba ±10ppm (1mL = 1mg Ba), LabChem™
Supplier: LabChem LC115501
Specifications
| Barium AA Standard | |
| Poly Bottle | |
| 99.82,0.18 | |
| 1g/mL | |
| BaCl2 | |
| BaCl2·2H2O | |
| Colorless | |
| WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
| barium(2+) dichloride | |
| 500 mL | |
| CHEBI:63317 | |
| Certified |
| 1000ppm (1mL = 1mg Ba) | |
| 10361-37-2 , 7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm Ba ±10 ppm (1mL = 1mg Ba) | |
| MFCD00003445 | |
| Soluble in water | |
| [Cl-].[Cl-].[Ba++] | |
| 208.23 | |
| 25204 | |
| 244.26 |
Chemical Identifiers
| 10361-37-2 | |
| 208.23 | |
| WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
| CHEBI:63317 | |
| [Cl-].[Cl-].[Ba++] |
| BaCl2 | |
| MFCD00003445 | |
| 25204 | |
| barium(2+) dichloride |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature