missing translation for 'onlineSavingsMsg'
Learn More

Barium AA Standard, 1000 ppm Ba in 3% HCl, Ricca Chemical

Supplier:  Ricca Chemical Company ABA1KH500

Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. ABA1KH500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

10361-37-2 , 7647-01-0 , 7732-18-5
208.227
25204
barium(2+);dichloride
BaCl2
WDIHJSXYQDMJHN-UHFFFAOYSA-L
CHEBI:63317
[Cl-].[Cl-].[Ba+2]

Specifications

Barium AA Standard
10361-37-2 , 7647-01-0 , 7732-18-5
94.0, 3.45, 0.15
1 mL = 1 mg Ba
WDIHJSXYQDMJHN-UHFFFAOYSA-L
barium(2+);dichloride
25204
Natural Poly Bottle
1,000ppm Ba
98.0, 3.59, 0.15
BaCl2
[Cl-].[Cl-].[Ba+2]
208.227
CHEBI:63317
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents