missing translation for 'onlineSavingsMsg'
Learn More

BAPTA tetramethyl ester, 99%

CAS: 125367-34-2 | C26H32N2O10 | 532.55 g/mol

Supplier:  Thermo Scientific Chemicals H6690703

 View more versions of this product

Catalog No. AAH6690703

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

BAPTA tetramethyl ester
White
MFCD00532664
bapta-tetramethyl ester, tetramethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, tetramethyl 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetate, methyl 2-2-2-2-bis 2-methoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-methoxy-2-oxoethyl amino acetate, 1,2-bis 2-bis 2-oxo-2-methoxyethyl amino phenoxy ethane, tetramethyl 1,2-bis 2-aminophenoxy ethane-n,n,n,n-tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n,n-tetraacetic acid, tetramethyl ester, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-2-methoxy-2-oxoethyl-, 1,1'-dimethyl ester, methyl 2-2-2-2-bis 2-methoxy-2-oxoethyl amino phenoxy ethoxy-n-2-methoxy-2-oxoethyl anilino acetate, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-2-methoxy-2-oxoethyl glycine 1,1'-dimethyl ester
COC(=O)CN(CC(=O)OC)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(=O)OC)CC(=O)OC
532.55
532.55
Powder
125367-34-2
C26H32N2O10
1 g
LCPLRKMZANZSAJ-UHFFFAOYSA-N
methyl 2-{[2-(2-{2-[bis(2-methoxy-2-oxoethyl)amino]phenoxy}ethoxy)phenyl](2-methoxy-2-oxoethyl)amino}acetate
3452402
99%

Chemical Identifiers

125367-34-2
532.55
LCPLRKMZANZSAJ-UHFFFAOYSA-N
3452402
COC(=O)CN(CC(=O)OC)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(=O)OC)CC(=O)OC
C26H32N2O10
MFCD00532664
bapta-tetramethyl ester, tetramethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, tetramethyl 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetate, methyl 2-2-2-2-bis 2-methoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-methoxy-2-oxoethyl amino acetate, 1,2-bis 2-bis 2-oxo-2-methoxyethyl amino phenoxy ethane, tetramethyl 1,2-bis 2-aminophenoxy ethane-n,n,n,n-tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n,n-tetraacetic acid, tetramethyl ester, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-2-methoxy-2-oxoethyl-, 1,1'-dimethyl ester, methyl 2-2-2-2-bis 2-methoxy-2-oxoethyl amino phenoxy ethoxy-n-2-methoxy-2-oxoethyl anilino acetate, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-2-methoxy-2-oxoethyl glycine 1,1'-dimethyl ester
methyl 2-{[2-(2-{2-[bis(2-methoxy-2-oxoethyl)amino]phenoxy}ethoxy)phenyl](2-methoxy-2-oxoethyl)amino}acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only