missing translation for 'onlineSavingsMsg'
Learn More

BAPTA tetramethyl ester, 99%

Catalog No. p-7057335
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66907MD 250 mg
AAH6690703 1 g
AAH6690706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66907MD Supplier Thermo Scientific Chemicals Supplier No. H66907MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 125367-34-2 | C26H32N2O10 | 532.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 125367-34-2
Molecular Formula C26H32N2O10
Molecular Weight (g/mol) 532.55
MDL Number MFCD00532664
InChI Key LCPLRKMZANZSAJ-UHFFFAOYSA-N
Synonym bapta-tetramethyl ester, tetramethyl 2,2',2,2'-ethane-1,2-diylbis oxy bis 2,1-phenylene bis azanetriyl tetraacetate, tetramethyl 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetate, methyl 2-2-2-2-bis 2-methoxy-2-oxoethyl amino phenoxy ethoxy phenyl 2-methoxy-2-oxoethyl amino acetate, 1,2-bis 2-bis 2-oxo-2-methoxyethyl amino phenoxy ethane, tetramethyl 1,2-bis 2-aminophenoxy ethane-n,n,n,n-tetraacetate, 1,2-bis 2-aminophenoxy ethane-n,n,n,n-tetraacetic acid, tetramethyl ester, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-2-methoxy-2-oxoethyl-, 1,1'-dimethyl ester, methyl 2-2-2-2-bis 2-methoxy-2-oxoethyl amino phenoxy ethoxy-n-2-methoxy-2-oxoethyl anilino acetate, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-2-methoxy-2-oxoethyl glycine 1,1'-dimethyl ester
PubChem CID 3452402
IUPAC Name methyl 2-{[2-(2-{2-[bis(2-methoxy-2-oxoethyl)amino]phenoxy}ethoxy)phenyl](2-methoxy-2-oxoethyl)amino}acetate
SMILES COC(=O)CN(CC(=O)OC)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(=O)OC)CC(=O)OC

Specifications

Color White
Quantity 250 mg
Formula Weight 532.55
Percent Purity 99%
Physical Form Powder
Chemical Name or Material BAPTA tetramethyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only