accessibility menu, dialog, popup

UserName

CH3OONOOCH3OONOOCH3OOCH3

CAS RN 125367-34-2

IUPAC Name: methyl 2-{[2-(2-{2-[bis(2-methoxy-2-oxoethyl)amino]phenoxy}ethoxy)phenyl](2-methoxy-2-oxoethyl)amino}acetate
Synonyms: methyl 2-{[2-(2-{2-[bis(2-methoxy-2-oxoethyl)amino]phenoxy}ethoxy)phenyl](2-methoxy-2-oxoethyl)amino}acetate
Molecular Weight (g/mol): 532.55
Molecular Formula: C26H32N2O10
InChi Key: LCPLRKMZANZSAJ-UHFFFAOYSA-N
SMILES: COC(=O)CN(CC(=O)OC)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(=O)OC)CC(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results