missing translation for 'onlineSavingsMsg'
Learn More

Bansyl Chloride 98.0+%, TCI America™

Catalog No. p-7118789
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09941G 1 g
B09945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09941G Supplier TCI America Supplier No. B09941G
Only null left

Chemical Identifiers

CAS 43040-76-2
Molecular Formula C18H24ClNO2S
Molecular Weight (g/mol) 353.91
MDL Number MFCD00059134
InChI Key PIZHBGYIKYGXBV-UHFFFAOYSA-N
Synonym 5-(Dibutylamino)naphthalene-1-sulfonyl Chloride
PubChem CID 191102
IUPAC Name 5-(dibutylamino)naphthalene-1-sulfonyl chloride
SMILES CCCCN(CCCC)C1=C2C=CC=C(C2=CC=C1)S(Cl)(=O)=O

Specifications

Quantity 1 g
UN Number 3265
Formula Weight 353.91
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Bansyl Chloride [N-Protecting Agent for Peptide Research]
TSCA No
Recommended Storage Refrigerator