missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bambuterol hydrochloride, Thermo Scientific™
$194.13 - $587.25
Chemical Identifiers
| CAS | 81732-46-9 |
|---|---|
| Molecular Formula | C18H29N3O5·HCl |
| Molecular Weight (g/mol) | 403.9 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC464910010
|
N/A | 1 g |
Each for $194.13
|
|
|||||
|
AC464910050
|
N/A | 5 g |
Each for $587.25
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 81732-46-9 | |
| C18H29N3O5·HCl | |
| 403.9 |
Specifications
| 81732-46-9 | |
| 403.9 | |
| Bambuterol hydrochloride |
| C18H29N3O5·HCl | |
| 403.9 |
RUO – Research Use Only