accessibility menu, dialog, popup

UserName

HClCH3NH3COOOONCH3CH3HOHNCH3H3CCH3

CAS RN 81732-46-9

IUPAC Name: hydrogen 3-[2-(tert-butylamino)-1-hydroxyethyl]-5-[(dimethylcarbamoyl)oxy]phenyl N,N-dimethylcarbamate chloride
Synonyms: bambuterol
Molecular Weight (g/mol): 403.9
Molecular Formula: C18H30ClN3O5
InChi Key: LBARATORRVNNQM-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CN(C)C(=O)OC1=CC(=CC(OC(=O)N(C)C)=C1)C(O)CNC(C)(C)C
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)