missing translation for 'onlineSavingsMsg'
Learn More

Bambuterol hydrochloride, Thermo Scientific Chemicals

Catalog No. AC464910050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464910050 5 g
AC464910010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464910050 Supplier Thermo Scientific Chemicals Supplier No. 464910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81732-46-9
Molecular Formula C18H30ClN3O5
Molecular Weight (g/mol) 403.90
MDL Number MFCD03427293
InChI Key LBARATORRVNNQM-UHFFFAOYNA-N
IUPAC Name hydrogen 3-[2-(tert-butylamino)-1-hydroxyethyl]-5-[(dimethylcarbamoyl)oxy]phenyl N,N-dimethylcarbamate chloride
SMILES [H+].[Cl-].CN(C)C(=O)OC1=CC(=CC(OC(=O)N(C)C)=C1)C(O)CNC(C)(C)C

Specifications

Melting Point 225.0°C to 229.0°C
Quantity 5 g
Formula Weight 403.9
Chemical Name or Material Bambuterol hydrochloride

RUO – Research Use Only