missing translation for 'onlineSavingsMsg'
Learn More

Bambuterol Hydrochloride 96.0+%, TCI America™

Catalog No. b45641g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B45641G 1 g
B45645G 5 g
2 options

Catalog No. B45641G

Supplier: TCI America B45641G

Only null left

Chemical Identifiers

CAS 81732-46-9
Molecular Formula C18H30ClN3O5
Molecular Weight (g/mol) 403.90
MDL Number MFCD03427293
InChI Key LBARATORRVNNQM-UHFFFAOYNA-N
Synonym bambuterol hydrochloride, bambuterol hcl, bambec tn, dimethylcarbmic acid 5-2-1,1-dimethylethyl amino-1-hydroxyethyl-1,3-phenylene ester, dsstox_cid_25515, dsstox_rid_80925, dsstox_gsid_45515, prestwick_777, bambuterolhydrochloride
PubChem CID 54765
ChEBI CHEBI:59167
IUPAC Name hydrogen 3-[2-(tert-butylamino)-1-hydroxyethyl]-5-[(dimethylcarbamoyl)oxy]phenyl N,N-dimethylcarbamate chloride
SMILES [H+].[Cl-].CN(C)C(=O)OC1=CC(=CC(OC(=O)N(C)C)=C1)C(O)CNC(C)(C)C

Specifications

Melting Point 225°C
Color White-Yellow
Quantity 1 g
Formula Weight 403.90
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bambuterol Hydrochloride