missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bacto™ Tryptose

Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties.
$494.79 - $7,777.50
Specifications
| Content And Storage | Store at 2°C to 25°C in cool, dry conditions in well-sealed containers. |
|---|---|
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Product Line | Bacto |
| Source | Animal Origin |
Description
Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptides of higher molecular weight suitable for long-chain amino acid requirements.
Features:
- Reported as beneficial for cell culture applications
- Animal origin (AO) nutritional supplement
Applications of Bacto Tryptose include:
- Tryptose is very useful for cultivation of streptococci, pneumococci, meningococci, and other fastidious organisms.
- Tryptose Peptone is often used as a biomass enhancer for recombinant Escherichia coli production.
Specifications
| Store at 2°C to 25°C in cool, dry conditions in well-sealed containers. | |
| Dehydrated Culture Media | |
| Animal Origin |
| Powder | |
| Bacto |