missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Tryptose
GREENER_CHOICE

Catalog No. 211709
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
211709 10 kg
DF0124172 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 211709 Supplier Gibco™ Supplier No. 211709

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties.

Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptides of higher molecular weight suitable for long-chain amino acid requirements.

Features:

  • Reported as beneficial for cell culture applications
  • Animal origin (AO) nutritional supplement

Applications of Bacto Tryptose include:

  • Tryptose is very useful for cultivation of streptococci, pneumococci, meningococci, and other fastidious organisms.
  • Tryptose Peptone is often used as a biomass enhancer for recombinant Escherichia coli production.

Specifications

Content And Storage Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Cell Culture
Product Line Bacto
Quantity 10 kg
Source Animal Origin