missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ TC Lactalbumin Hydrolysate

An animal origin peptone from the enzymatically hydrolyzed protein portion of milk whey.

$237.93 - $3,413.82

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Mammalian, bacterial, insect cell types, recombinant proteins and animal health vaccines
Product Line Bacto™
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
259961
View Documents
Gibco™
259961
10 kg N/A
Only null left
 
DF5996-17-0
View Documents
Gibco™
259962
500 g N/A
Only null left
 
Description

Description

This supplement is a mixture of peptides, amino acids, and carbohydrates, both simple and complex. Customers have successfully used Gibco™ Bacto™ TC Lactalbumin Hydrolysate to support the growth of many mammalian, bacterial, and insect cell types, and it has been used to produce both recombinant proteins and animal health vaccines. Customers have also used this supplement as an amino acid supplement for mammalian cell culture media.
Specifications

Specifications

Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Dehydrated Culture Media
Bacto™
Powder
Mammalian, bacterial, insect cell types, recombinant proteins and animal health vaccines
Animal Origin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'