missing translation for 'onlineSavingsMsg'
Learn More

Cytiva bacteria genomicPrep Mini Spin Kit

Designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria

$426.00 - $1926.79

Specifications

Description illustra bacteria genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria.
For Use With (Application) For the rapid extraction and purification of high-quality genomic DNA from gram-positive and gram-negative bacteriaPurified gDNA is suitable for molecular biology applications including cloning, restriction enzyme digestion, PCR and genotyping
Product Type illustra bacteria genomicPrep Mini Spin Kit
Purity 1.7
Sample Volume 2 x 109 cells [A600 = 2.0]
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity  
Catalog Number Mfr. No. No. of Reactions Price Quantity  
28-904-258
View Documents
Cytiva
28904258
50 Preps
Each for $426.00
Only null left
 
45-001-494
View Documents
Cytiva
28904259
250 Preps
Each for $1,926.79
Only null left
 
Description

Description

The illustra Bacteria genomicPrep Mini Spin Kit can be used with a wide range of bacterial strains such as DH5a, TOP10, JM109, Bacillus subtilis etc. The kit produces highly pure and intact genomic DNA with OD260/280 values of 1.75-1.85. Use different elution volumes to isolate genomic DNA at concentrations suitable for most downstream applications including restriction enzyme digestion, RT-PCR, and long PCR.

The method uses a lysis solution in combination with proteinase K to gently release genomic DNA into solution from bacteria cells. The DNA is deproteinated in the extraction solution and the genomic DNA is then bound onto a silica column in the presence of chaotropic solution. Contaminants are removed during wash steps and the purifed DNA is eluted with prewarmed TE solution.

  • Total DNA extraction time under one hour with reduced lysis time
  • Can be used to purify 4 to 8μg of genomic DNA from 2 × 109 bacteria cells with input amounts ranging from 1 × 109 to 4 × 109 bacteria cells
  • Spin columns prepacked with a silica membrane—provide excellent reproducibility and consistency
  • Validated in several downstream applications including RT PCR, long PCR, and restriction digest
  • Color-coded caps for ease of use

Specifications

Specifications

illustra bacteria genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria.
illustra bacteria genomicPrep Mini Spin Kit
2 x 109 cells [A600 = 2.0]
For the rapid extraction and purification of high-quality genomic DNA from gram-positive and gram-negative bacteriaPurified gDNA is suitable for molecular biology applications including cloning, restriction enzyme digestion, PCR and genotyping
1.7
40 min [Gram-negaitive Bacteria], 55 min [Gram-positive Bacteria]
Videos
SDS
Documents

Documents