missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ b-Cyclodextrin

Catalog No. 31811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31811G Supplier Tocris Bioscience™ Supplier No. 3181/1G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cyclic complexing agent

β-Cyclodextrin (β-CD) is a cyclic oligosaccharide comprising seven glucose subunits linked by α-1,4-glycosidic bonds, forming a cage-like supramolecular structure with a hydrophobic interior and a hydrophilic exterior. β-Cyclodextrins can form inclusion complexes with a diverse range of hydrophobic compounds. These complexes increase the aqueous solubility, dissolution rate, and bioavailability of the guest compounds.

Specifications

CAS 7585-39-9
Quantity 1 g
Target Buffers, Chelators and Reagents
Molecular Formula C42H70O35
Purity 98%