missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ B-27™ Supplement, minus insulin
B-27™ Supplement, minus insulin
$296.00 - $2,715.00
Specifications
| Cell Type | Neural Cells |
|---|---|
| Classification | Serum-free |
| Concentration | 50 X |
| Culture Type | Mammalian Cell Culture |
| Form | Liquid |
Description
B-27™ Supplement Minus Insulin is the complete B-27 Supplement without insulin. It is used to support growth of CNS neurons and other cells in applications where the presence of insulin causes interference.For neural studies that require the absence of insulin, glucose, and sodium pyruvate, we recommend Neurobasal™-A without D-glucose and sodium pyruvate (Cat. No. 0050128DJ) as a basal medium for use in conjunction with this product.
For other applications, we provide alternative formulations of B-27 Supplement.
Specifications
| Neural Cells | |
| 50 X | |
| Liquid | |
| Serum-Free | |
| Dry Ice | |
| Endotoxin, Performance, Sterility, Mycoplasma |
| Serum-free | |
| Mammalian Cell Culture | |
| Cell Culture Supplement | |
| B-27 | |
| Serum-Free |
For Research Use Only. Not for use in diagnostic procedures.