missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Aztreonam | |
| 227°C | |
| C13H17N5O8S2 | |
| 500 mg | |
| WZPBZJONDBGPKJ-PSGLRMFWSA-N | |
| 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid | |
| 56842327 | |
| ≥98.0% (HPLC) |
| 78110-38-0 | |
| White | |
| MFCD00072145 | |
| trans-2-1-2-aminothiazol-4-yl-2-2-methyl-4-oxo-1-sulphoazetidin-3-yl amino-2-oxoethylidene amino oxy-2-methylpropionic acid, 2-1-2-amino-4-thiazolyl-2-2r-2alpha-methyl-4-oxo-1-sulfo-3beta-azetidinyl amino-2-oxoethylidene amino oxy-2-methylpropanoic acid | |
| CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N | |
| 435.426 | |
| 435.43 | |
| Crystalline Powder |
Chemical Identifiers
| 78110-38-0 | |
| 435.426 | |
| WZPBZJONDBGPKJ-PSGLRMFWSA-N | |
| 56842327 | |
| CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N |
| C13H17N5O8S2 | |
| MFCD00072145 | |
| trans-2-1-2-aminothiazol-4-yl-2-2-methyl-4-oxo-1-sulphoazetidin-3-yl amino-2-oxoethylidene amino oxy-2-methylpropionic acid, 2-1-2-amino-4-thiazolyl-2-2r-2alpha-methyl-4-oxo-1-sulfo-3beta-azetidinyl amino-2-oxoethylidene amino oxy-2-methylpropanoic acid | |
| 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid |
Safety and Handling
RTECSNumber : UA2451400
TSCA : No