missing translation for 'onlineSavingsMsg'
Learn More

Aztreonam 98.0+%, TCI America™

Catalog No. p-7117926
Change view
Click to view available options
Quantity:
5 g
500 mg
Catalog No. Quantity
A2466500MG 500 mg
A24665G 5 g
2 options

Catalog No. A2466500MG

Supplier: TCI America A2466500MG

Only null left

Chemical Identifiers

CAS 78110-38-0
Molecular Formula C13H17N5O8S2
Molecular Weight (g/mol) 435.426
MDL Number MFCD00072145
InChI Key WZPBZJONDBGPKJ-PSGLRMFWSA-N
Synonym trans-2-1-2-aminothiazol-4-yl-2-2-methyl-4-oxo-1-sulphoazetidin-3-yl amino-2-oxoethylidene amino oxy-2-methylpropionic acid, 2-1-2-amino-4-thiazolyl-2-2r-2alpha-methyl-4-oxo-1-sulfo-3beta-azetidinyl amino-2-oxoethylidene amino oxy-2-methylpropanoic acid
PubChem CID 56842327
IUPAC Name 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
SMILES CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N

Specifications

Melting Point 227°C
Color White
Quantity 500 mg
Formula Weight 435.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Aztreonam
RTECSNumber UA2451400
TSCA No