missing translation for 'onlineSavingsMsg'
Learn More

Azetidine-3-carboxylic acid, 98+%

CAS: 36476-78-5 | C4H7NO2 | 101.11 g/mol

Supplier:  Thermo Scientific Chemicals H5229403

 View more versions of this product

Catalog No. AAH5229403


Only null left
Explore available promotions

Description

Description

Azetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Azetidine-3-carboxylic acid
∽250°C (decomposition)
MFCD00191763
3-azetidinecarboxylic acid, 3-azetanecarboxylic acid, 3-carboxyazetidine, caswell no. 063c, h-aze 3-oh, azetane-3-carboxylic acid, epa pesticide chemical code 128830, 3-azetidine carboxylic acid, l-azetidine-3-carboxylic acid, azetidine-3-carboxylicacid
GFZWHAAOIVMHOI-UHFFFAOYSA-N
azetidine-3-carboxylic acid
93192
≥98%
36476-78-5
C4H7NO2
1 g
Soluble in water.
OC(=O)C1CNC1
101.11
101.11

Chemical Identifiers

36476-78-5
101.11
GFZWHAAOIVMHOI-UHFFFAOYSA-N
93192
OC(=O)C1CNC1
C4H7NO2
MFCD00191763
3-azetidinecarboxylic acid, 3-azetanecarboxylic acid, 3-carboxyazetidine, caswell no. 063c, h-aze 3-oh, azetane-3-carboxylic acid, epa pesticide chemical code 128830, 3-azetidine carboxylic acid, l-azetidine-3-carboxylic acid, azetidine-3-carboxylicacid
azetidine-3-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CM4310600

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only