accessibility menu, dialog, popup

UserName

HOONH

CAS RN 36476-78-5

IUPAC Name: azetidine-3-carboxylic acid
Synonyms: 3-azetidinecarboxylic acid
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: GFZWHAAOIVMHOI-UHFFFAOYSA-N
SMILES: OC(=O)C1CNC1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)