missing translation for 'onlineSavingsMsg'
Learn More

Azetidine-3-carboxylic acid, 98+%

Catalog No. p-7034607
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52294MD 250 mg
AAH5229403 1 g
AAH5229406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52294MD Supplier Thermo Scientific Chemicals Supplier No. H52294MD

CAS: 36476-78-5 | C4H7NO2 | 101.11 g/mol

Azetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 36476-78-5
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00191763
InChI Key GFZWHAAOIVMHOI-UHFFFAOYSA-N
Synonym 3-azetidinecarboxylic acid, 3-azetanecarboxylic acid, 3-carboxyazetidine, caswell no. 063c, h-aze 3-oh, azetane-3-carboxylic acid, epa pesticide chemical code 128830, 3-azetidine carboxylic acid, l-azetidine-3-carboxylic acid, azetidine-3-carboxylicacid
PubChem CID 93192
IUPAC Name azetidine-3-carboxylic acid
SMILES OC(=O)C1CNC1

Specifications

Melting Point ∽250°C (decomposition)
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 101.11
Percent Purity ≥98%
Chemical Name or Material Azetidine-3-carboxylic acid
RTECSNumber CM4310600
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only