missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ AZD 4547
Description
AZD 4547 is a potent and selective FGFR1, FGFR3 and FGFR2 tyrosine kinase inhibitor (IC50 values are 0.2 1.8, and 2.5 nM, respectively). It shows selectivity for FGFR across a range of unrelated tyrosine and serine/threonine kinases, including IGFR (>2,900-fold), CDK2 (>50,000-fold), and p38 (>50,000-fold). In vitro, it has antiproliferative effects on tumor cell lines with deregulated FGFR expression. In vivo, AZD 4547 exhibits dose-dependent inhibition of tumor growth in an FGFR-driven human tumor xenograft mice model. It suppresses osteoclastogenesis and tumor-induced osteolysis in an orthotopic breast cancer bone metastasis mouse model. AZD 4547 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | rel-N-[5-[2-(3,5-Dimethoxyphenyl)ethyl]-1H-pyrazol-3-yl]-4-[(3R,5S)-3,5-dimethyl-1-piperazinyl]benzamide |
| CAS | 1035270-39-3 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | FGF Receptor Inhibitors |
| Molecular Formula | C26H33N5O3 |