missing translation for 'onlineSavingsMsg'
Learn More

Azathioprine 98.0+%, TCI America™

Supplier:  TCI America A20695G

 View more versions of this product

Catalog No. A20695G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Azathioprine
244°C
MFCD00069203
azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]
277.262
CHEBI:2948
≥98.0% (HPLC,T)
446-86-6
C9H7N7O2S
5 g
LMEKQMALGUDUQG-UHFFFAOYSA-N
6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
2265
277.26
Crystalline Powder

Chemical Identifiers

446-86-6
277.262
LMEKQMALGUDUQG-UHFFFAOYSA-N
2265
6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
C9H7N7O2S
MFCD00069203
azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
CHEBI:2948
CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1448

RTECSNumber : UO8925000

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions