missing translation for 'onlineSavingsMsg'
Learn More

Azathioprine 98.0+%, TCI America™

Catalog No. p-7117711
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20695G 5 g
A206925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20695G Supplier TCI America Supplier No. A20695G
Only null left

Chemical Identifiers

CAS 446-86-6
Molecular Formula C9H7N7O2S
Molecular Weight (g/mol) 277.262
MDL Number MFCD00069203
InChI Key LMEKQMALGUDUQG-UHFFFAOYSA-N
Synonym azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
PubChem CID 2265
ChEBI CHEBI:2948
IUPAC Name 6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
SMILES CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]

Specifications

Melting Point 244°C
Quantity 5 g
Formula Weight 277.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Azathioprine
EINECSNumber (9)-1448
RTECSNumber UO8925000
TSCA No
Recommended Storage Freezer