missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Azasetron hydrochloride

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

$325.95 - $893.37

Chemical Identifiers

CAS 123040-16-4
Molecular Formula C17H21Cl2N3O3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AC465070010
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
465070010
1 g
Each for $893.37
Only null left
 
AC465072500
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
465072500
250 mg
Each for $325.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123040-16-4
C17H21Cl2N3O3

Specifications

123040-16-4
C17H21Cl2N3O3
Powder
≥97.5% (HPLC)
[H+].[Cl-].CN1C(=O)COC2=C1C=C(Cl)C=C2C(=O)NC1CN2CCC1CC2
386.27
1 g
386.27
Reagent
DBMKBKPJYAHLQP-UHFFFAOYNA-N
hydrogen N-{1-azabicyclo[2.2.2]octan-3-yl}-6-chloro-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxamide chloride
Azasetron hydrochloride
Videos
SDS
Documents

Documents

RUO – Research Use Only