missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Azasetron hydrochloride

Catalog No. AC465070010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465070010 1 g
AC465072500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465070010 Supplier Thermo Scientific Chemicals Supplier No. 465070010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123040-16-4
Molecular Formula C17H21Cl2N3O3

Specifications

Quantity 1 g
Formula Weight 386.27
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
InChI Key DBMKBKPJYAHLQP-UHFFFAOYNA-N
SMILES [H+].[Cl-].CN1C(=O)COC2=C1C=C(Cl)C=C2C(=O)NC1CN2CCC1CC2
IUPAC Name hydrogen N-{1-azabicyclo[2.2.2]octan-3-yl}-6-chloro-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxamide chloride
Molecular Weight (g/mol) 386.27
Chemical Name or Material Azasetron hydrochloride

RUO – Research Use Only