missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZ1 USP25/28 inhibitor

Catalog No. 784510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
784510 10 mg
784550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 784510 Supplier Tocris Bioscience™ Supplier No. 7845/10

Selective USP25 and USP28 inhibitor

AZ1 USP25/28 inhibitor is a selective inhibitor of USP28 (IC50 = 0.6 μM, Kd= 0.2 - 3.7 μM) and USP25 (IC50 = 0.7 μM). AZ1 USP25/28 inhibitor exhibits selectivity for USP25 and USP28 over other DUB family members. The compound dose-dependently reduces c-Myc levels in colon carcinoma cells in vitro and induces cell death (EC50 values are 18 - 20 μM). In a mouse Alzheimer's disease model, AZ1 USP25/28 inhibitor reduces amyloid burden, attenuates microglial activation and improves synaptic and cognitive function.

Specifications

Chemical Name or Material 2-[[[5-Bromo-2-[[4-fluoro-3-(trifluoromethyl)phenyl]methoxy]phenyl]methyl]amino]ethanol
CAS 2165322-94-9
Quantity 10 mg
Target USP28
Molecular Formula C17H16BrF4NO2
Purity 0.98