missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZ 12799734
Description
AZ 12799734 is a potent inhibitor of transforming growth factor-β type I receptor (TGF-βRI; ALK5) (IC50 = 17 nM in cellular inhibition assay and 47 nM in TGF-β-induced luciferase activity assay). Also inhibits ALK4, ALK6 and ALK7 at concentrations under 10 μM. Inhibits BMP signaling, phosphorylation of SMAD1 and SMAD2, and TGF-β-induced epithelial cell migration in vitro. Displays cardiotoxicity in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 4-[[4-[(2.6-Dimethyl-3-pyridinyl)oxy]-2-pyridinyl]amino]benzenesulfonamide |
| CAS | 1117684-36-2 |
| Quantity | 50 mg |
| Molecular Formula | C18H18N4O3S |
| Purity | 98% |