Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing with Conical Bottoms
Self-standing tubes
Supplier:  Axygen™ ST150SSCS
 View more versions of this product
Catalog No. 14-222-675
Only null left
Description
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
Specifications
Specifications
RNase, DNase, pyrogen-free
Clear
Bag
Not Included
20,000 x g
1.5 mL
Polypropylene
Sterile
Conical
Product Suggestions
Product Suggestions