missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ IMAG Handheld Magnetic Beads Separation Devices
GREENER_CHOICE

Catalog No. 14223402
Change view
Click to view available options
Format:
96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
14223402 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14223402 Supplier Axygen™ Supplier No. IMAG96P

Transform low throughput to high throughput separation by a flip of a tube or microplate

The use of magnetic separation devices (MSDs) are essential in any paramagnetic beads-based purification process. Traditionally, MSDs are not optimized for manual use and most require electrically powered liquid handling systems. IMAG MSDs are available in tube or microplate format and are designed to simplify the manual processing of magnetic bead separations, including nucleic acid purification and clean-up, cell-based assays, and antibody and protein purifications.

  • IMAG MSDs accommodate single tube or 96-well microplate formats
  • Fast separation time

        - Strong magnets enable less than 30-second separation times

        - Holds tubes or microplate enabling fast wash steps

  • Manual operation enables low or high throughput

Specifications

Format 96-well Plate
Isolation Technology Magnetic Bead
For Use With (Equipment) 96 Microplates
For Use With (Application) Nucleic Acid purification and clean up, cell based assays, Antibody and Protein purification
Includes Strong magnets providing less than 30 sec. separation time
Product Type Magnetic Separation Device
Quantity Each