missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ AxyPrep™ Mag Plasmid Kits

Catalog No. 14223166
Change view
Click to view available options
No. of Reactions:
384 Preps
96 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
14223166 96 Preps
14223167 384 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14223166 Supplier Axygen™ Supplier No. MAGPS

Utilizes magnetic bead technology for high-throughput purification of plasmid DNA from E.coli cells

The AxyPrep MAG Plasmid purification kit utilizes a magnetic beads-based technology for high throughput purification of plasmid DNA from Escherichia coli cells. AxyPrep MAG Plasmid Kit can also be used with fosmid and BAC vector-based constructs ranging from 5 Kb to 150 Kb. The system consists of steps of pelleting cells, resuspension, lysis, neutralization, DNA binding, ethanol washing, and plasmid elution. The protocol is automation-compatible.

Specifications

Format 96-well Plate, Tube
For Use With (Application) DNA sequencing, PCR amplification, restriction Enzyme digestion
No. of Reactions 96 Preps
Product Type Kit