Learn More
Axygen™ Axyprep™ MAG Plant Genomic DNA Extraction Kit
Magnetic beads based purification system allows rapid and reliable isolation of genomic DNA from a wide variety of plant samples. Up to 96 samples (50 mg fresh tissue per sample, or 15 mg of dried tissue per sample) can be processed in less than an hour.
Supplier: Axygen™ MAGPLANTGDNAS
Description
The AxyPrep MAG Plant gDNA Extraction Kit is a magnetic beads-based purification system that allows rapid and reliable isolation of high quality gDNA from a wide variety of plant samples. Up to 96 samples (50 mg fresh tissue per sample or 15 mg of dried tissue per sample) can be processed in less than one hour. The AxyPrep MAG Plant gDNA Extraction Kit efficiently eliminates polysaccharides, phenolic compounds and enzyme inhibitors with no organic extractions. This kit is adaptable for use on major liquid handling workstations for high throughput processing.
- Simplified extraction process
- Non-alcohol-based purification
- Compatible with genomics downstream applications
- High quality, high-yield DNA
- Manual and automation friendly
Specifications
| DNA extraction and purification | |
| MAG-A1 Particles 2.2 mL; PCL Buffer 60 mL; PDB Buffer 60 mL; HSW Buffer 44 mL; MB Elution Buffer 15 mL; Protocol Manual 1 | |
| 96 Preps | |
| MAG Plant Genomic DNA Extraction Kit |