missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ AxyPrep Mag™ DyeClean Kits
Description
The AxyPrep Mag DyeClean Kit utilizes a unique paramagnetic bead-based sequencing purification system that is optimized for the removal of excess dye terminator from Sanger sequencing reactions. The process consists of a selective binding of DNA to the beads, a wash step for removing nucleotides, primers, and non-targeted amplicons, and elution of pure DNA. Through use of this kit you can achieve Rapid and reliable clean-up of sequencing reactions while maintaining high QV20+ scores. This kit can be used for manual or automated processes.
- High DNA recovery yields, long sequence readings
- Suitable for the clean-up of sequencing reactions of different Sanger sequencing based platforms - ABI Prism 3730, 3730xl, 3700, 3130, 31030xl and 3100 - GE Healthcare MegaBACE
- Simple and easy sequencing reaction clean-up process: 25 minutes/96 samples
- Scalable: Tube, 96 and 384 well plate formats
- No centrifugation or filtration required
Specifications
Specifications
| Format | 96- or 384-well Plate |
| For Use With (Equipment) | ABI PRISM 3730, 3730xl, 3700, 3130, 31030xl and 3100, MegaBACE |
| For Use With (Application) | Removes excess dye terminator chemistries |
| No. of Reactions | 500 to 700 Preps |
| Product Type | Kit |
| Quantity | 5 mL |