missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 0.5 mL Self Standing Screw Cap Tubes
Features O-ring manufactured for resistant to many commonly-used laboratory chemicals
$325.63 - $2726.04
Specifications
| Certifications/Compliance | RNase, DNase, pyrogen-free |
|---|---|
| Closure Material | Polypropylene |
| Capacity (Metric) | 0.5 mL |
| Material | Polypropylene |
| Format | Non-looped |
| Catalog Number | Mfr. No. | Sterility | Closure Color | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sterility | Closure Color | Price | Quantity | ||||
|
14-222-534
|
Axygen™
SCT050SSAS |
Sterile | Assorted |
Case of 4000 Each for $2,726.04
|
|
||||
|
14-222-539
|
Axygen™
SCT050SSC |
Non-sterile | Clear |
Case of 4000 Each for $2,166.56
|
|
||||
|
14-222-540
|
Axygen™
SCT050SSCS |
Sterile | Clear |
Pack of 500 Each for $325.63
Case of 8 Pack for $2,605.04
|
|
||||
|
14-222-541
|
Axygen™
SCT050SSG |
Non-sterile | Green |
Case of 4000 Each for $2,498.06
|
|
||||
|
14-222-544
|
Axygen™
SCT050SSGS |
Sterile | Green |
Case of 4000 Each for $2,345.83
|
|
||||
|
14-222-546
|
Axygen™
SCT050SSOS |
Sterile | Orange |
Case of 4000 Each for $2,487.22
|
|
||||
|
14-222-548
|
Axygen™
SCT050SSR |
Non-sterile | Red |
Case of 4000 Each for $2,416.60
|
|
||||
|
14-222-549
|
Axygen™
SCT050SSRS |
Sterile | Red |
Case of 4000 Each for $2,414.79
|
|
||||
|
14-222-555
|
Axygen™
SCT050SSYS |
Sterile | Yellow |
Case of 4000 Each for $2,414.79
|
|
||||
Description
For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
- Available as presterilized and nonsterile tubes
- Nonsterile cap tubes have unattached, non-looped caps with O-rings
- Presterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
- Wide range of colored tubes for easy sample identification
Specifications
| RNase, DNase, pyrogen-free | |
| 0.5 mL | |
| Non-looped | |
| Screw Cap | |
| 20,000 x g |
| Polypropylene | |
| Polypropylene | |
| O-ring | |
| Conical |