Learn More
Axygen™ 0.5 mL Self Standing Screw Cap Tubes
Features O-ring manufactured for resistant to many commonly-used laboratory chemicals
Supplier:  Axygen™ SCT050SSC
 View more versions of this product
Catalog No. 14-222-539
Only null left
Explore available promotions
Description
Description
For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile tubes
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Wide range of colored tubes for easy sample identification
Specifications
Specifications
RNase, DNase, pyrogen-free
Polypropylene
Polypropylene
O-ring
No
Conical
Clear
0.5 mL
Non-looped
Non-sterile
Screw Cap
20,000 x g
Product Suggestions
Product Suggestions