missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Axitinib

Catalog No. 435010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
435010 10 mg
435050 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 435010 Supplier Tocris Bioscience™ Supplier No. 4350/10*

Potent VEGFR-1, -2 and -3 inhibitor

Axitinib is a potent inhibitor of VEGFR-2, -3, and -1 (IC50 values are 0.2, 0.1-0.3, and 1.2 nM respectively). Axitinib exhibits minimal activity against a panel of ∼100 protein kinases. Axitinib inhibits angiogenesis and vascular permeability. Also a high affinity BCR-ABL1 (T315I) inhibitor (Ki = 149 pM for autophosphorylated ABL1 (T315I)). Axitinib blocks proliferation of Ba/F3 cells expressing BCR-ABL1 (T315I). Axitinib (10 mg/kg) decreases T-cell exhaustion and tumor and T-cell hypoxia and improves survival in mice in vivo. Orally available.

Chemical Identifiers

CAS 319460-85-0
Molecular Formula C22H18N4OS

Specifications

Quantity 10 mg
Formula Weight 386.47
Purity >99%
Molecular Weight (g/mol) 386.473
Chemical Name or Material Axitinib
Recommended Storage Store at +4°C