missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Avermectin, ACROS Organics™

$277.35 - $923.91

Specifications

Chemical Name or Material Avermectin
CAS 65195-55-3
Molecular Formula C48 H72 O14
Formula Weight 873.09
Physical Form Crystalline Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC468392500
View Documents
Thermo Scientific™
468392500
250 mg
Each for $277.35
Only null left
 
AC468390010
View Documents
Thermo Scientific™
468390010
1 g
Each for $923.91
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Avermectin
C48 H72 O14
Crystalline Powder
65195-55-3
873.09
White to Light-yellow
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'